6-Chloro-2,3-difluorobenzotrifluoride
Supplier: Apollo Scientific
PC450023-1G PC450023-250MG
APOSPC450023-1GEA 880 EUR
APOSPC450023-1G APOSPC450023-250MG
6-Chloro-2,3-difluorobenzotrifluoride
6-Chloro-2,3-difluorobenzotrifluoride
Formula: C₇H₂ClF₅
MW: 216.536 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12922577
CAS Number: 1208076-15-6