Perfluoro-2-propanesulphonyl fluoride 97%
Supplier: Apollo Scientific
Danger
PC450051-1G PC450051-5G
APOSPC450051-1GEA 354 EUR
APOSPC450051-1G APOSPC450051-5G
Perfluoro-2-propanesulphonyl fluoride 97%
Perfluoro-2-propanesulphonyl fluoride
Formula: C₃F₈O₂S
MW: 252.083 g/mol
Boiling Pt: 34…40 °C
Melting Pt: –59 °C
Storage Temperature: Ambient
MDL Number: MFCD20489409
CAS Number: 14856-91-8
UN: 3265
ADR: 8,II