4-Bromo-1,1-difluorocyclohexane
Supplier: Apollo Scientific
Warning
PC450063-1G PC450063-250MG
APOSPC450063-1GEA 765 EUR
APOSPC450063-1G APOSPC450063-250MG
4-Bromo-1,1-difluorocyclohexane
4-Bromo-1,1-difluorocyclohexane
Formula: C₆H₉BrF₂
MW: 199.036 g/mol
Boiling Pt: 49…50 °C (11 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD13189572
CAS Number: 1196156-51-0