1,1-Dibromo-3,3-difluoroacetone 97%
Supplier: Apollo Scientific
Warning
PC450064-1G PC450064-250MG PC450064-5G
APOSPC450064-1GEA 292 EUR
APOSPC450064-1G APOSPC450064-250MG APOSPC450064-5G
1,1-Dibromo-3,3-difluoroacetone 97%
1,1-Dibromo-3,3-difluoroacetone
Formula: C₃H₂Br₂F₂O
MW: 251.852 g/mol
Boiling Pt: 64…66 °C
Storage Temperature: Ambient
MDL Number: MFCD19442051
CAS Number: 1309602-53-6