2-Bromo-2,2-difluoro-1-phenylethanol
Supplier: Apollo Scientific
PC450099-1G PC450099-250MG
APOSPC450099-1GEA 1230 EUR
APOSPC450099-1G APOSPC450099-250MG
2-Bromo-2,2-difluoro-1-phenylethanol
2-Bromo-2,2-difluoro-1-phenylethanol
Formula: C₈H₇BrF₂O
MW: 237.041 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18074357
CAS Number: 74492-28-7