2-(Trifluoromethyl)-1,3-benzothiazole
Supplier: Apollo Scientific
PC450127-1G PC450127-5G
APOSPC450127-1GEA 139 EUR
APOSPC450127-1G APOSPC450127-5G
2-(Trifluoromethyl)-1,3-benzothiazole
2-(Trifluoromethyl)-1,3-benzothiazole
Formula: C₈H₄F₃NS
MW: 203.184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00065209
CAS Number: 14468-40-7