(S)-1-(2-Fluoro-4-methoxyphenyl)ethylamine hydrochloride
Supplier: Apollo Scientific
Synonyms: (S)-1-(2-Fluoro-4-methoxyphenyl)ethanamine hydrochloride
PC450132-1G PC450132-250MG
APOSPC450132-1GEA 579 EUR
APOSPC450132-1G APOSPC450132-250MG
(S)-1-(2-Fluoro-4-methoxyphenyl)ethylamine hydrochloride
(S)-1-(2-Fluoro-4-methoxyphenyl)ethylamine hydrochloride
Formula: C₉H₁₂FNO·ClH
MW: 205.657 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16294167
CAS Number: 1149383-12-9