4-Cyano-3-(trifluoromethyl)trifluoroacetanilide
Supplier: Apollo Scientific
Synonyms: N-(4-Cyano-3-(trifluoromethyl)phenyl)-2,2,2-trifluoroacetamide
PC450133-1G PC450133-250MG
APOSPC450133-1GEA 369 EUR
APOSPC450133-1G APOSPC450133-250MG
4-Cyano-3-(trifluoromethyl)trifluoroacetanilide
4-Cyano-3-(trifluoromethyl)trifluoroacetanilide
Formula: C₁₀H₄F₆N₂O
Storage Temperature: Ambient
MDL Number: MFCD18074383
CAS Number: 1155800-45-5