1,1'-Oxybis[4-(tridecafluorohexyl)benzene
Supplier: Apollo Scientific
PC450140-100MG PC450140-1G PC450140-250MG
APOSPC450140-100MGEA 139 EUR
APOSPC450140-100MG APOSPC450140-1G APOSPC450140-250MG
1,1'-Oxybis[4-(tridecafluorohexyl)benzene
1,1'-Oxybis[4-(tridecafluorohexyl)benzene
Formula: C₂₄H₈F₂₆O
MW: 806.278 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24842590
CAS Number: 151707-03-8