Perfluoro(4-methyl-2-pentenoic acid)
Supplier: Apollo Scientific
Danger
Synonyms: (E)-Perfluoro(4-methylpent-2-enoic acid), Hexafluoro-4-(trifluoromethyl)pent-2-enoic acid, Nonafluoro(4-methylpent-2-enoic acid), 4-(Trifluoromethyl)hexafluoropent-2-enoic acid
PC450153-1G
APOSPC450153-1GEA 87.3 EUR
APOSPC450153-1G
Perfluoro(4-methyl-2-pentenoic acid)
Perfluoro(4-methyl-2-pentenoic acid)
Formula: C₆HF₉O₂
MW: 276.06 g/mol
Melting Pt: 173…174 °C
Storage Temperature: Ambient
MDL Number: MFCD00153286
CAS Number: 239795-58-5
UN: 3265
ADR: 8,III