5,6-Dimethyl-2-(trifluoromethyl)-1H-benzimidazole
Supplier: Apollo Scientific
PC450156-1G PC450156-5G PC450156-250MG
APOSPC450156-1GEA 323 EUR
APOSPC450156-1G APOSPC450156-5G APOSPC450156-250MG
5,6-Dimethyl-2-(trifluoromethyl)-1H-benzimidazole
5,6-Dimethyl-2-(trifluoromethyl)-1H-benzimidazole
Formula: C₁₀H₉F₃N₂
MW: 214.187 g/mol
Melting Pt: 235…238 °C
Storage Temperature: Ambient
MDL Number: MFCD00022676
CAS Number: 1799-79-7
UN: 2811
ADR: 6.1,III