Tris(2,2-difluoroethyl)borate
Supplier: Apollo Scientific
Warning
PC450157-1G PC450157-250MG PC450157-5G PC450157-100MG
APOSPC450157-1GEA 737 EUR
APOSPC450157-1G APOSPC450157-250MG APOSPC450157-5G APOSPC450157-100MG
Tris(2,2-difluoroethyl)borate
Tris(2,2-difluoroethyl)borate
Formula: C₆H₉BF₆O₃
MW: 253.935 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19442172
CAS Number: 1396338-09-2
UN: 1993
ADR: 3,III