(S)-1-(3,4-Difluorophenyl)ethanol
Supplier: Apollo Scientific
PC450161-1G PC450161-5G
APOSPC450161-1GEA 430 EUR
APOSPC450161-1G APOSPC450161-5G
(S)-1-(3,4-Difluorophenyl)ethanol
(S)-1-(3,4-Difluorophenyl)ethanol
Formula: C₈H₈F₂O
MW: 158.145 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09863749
CAS Number: 126534-40-5