1-Allyl-2-(trifluoromethyl)benzimidazole
Supplier: Apollo Scientific
PC450171-1G PC450171-250MG
APOSPC450171-1GEA 333 EUR
APOSPC450171-1G APOSPC450171-250MG
1-Allyl-2-(trifluoromethyl)benzimidazole
1-Allyl-2-(trifluoromethyl)benzimidazole
Formula: C₁₁H₉F₃N₂
MW: 226.198 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02102641
CAS Number: 139591-03-0