1-(3-Fluorophenyl)-2,2,2-trifluoroethanol
Supplier: Apollo Scientific
Synonyms: 2,2,2-Trifluoro-1-(3-fluorophenyl)ethanol
PC450180-1G PC450180-5G
APOSPC450180-1GEA 139 EUR
APOSPC450180-1G APOSPC450180-5G
1-(3-Fluorophenyl)-2,2,2-trifluoroethanol
1-(3-Fluorophenyl)-2,2,2-trifluoroethanol
Formula: C₈H₆F₄O
MW: 194.126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16067876
CAS Number: 81577-10-8