2-[2-(2,2,3,3-Tetrafluoropropoxy)ethoxy]ethan-1-ol
Supplier: Apollo Scientific
PC450182-1G PC450182-250MG PC450182-5G
APOSPC450182-1GEA 168 EUR
APOSPC450182-1G APOSPC450182-250MG APOSPC450182-5G
2-[2-(2,2,3,3-Tetrafluoropropoxy)ethoxy]ethan-1-ol
2-[2-(2,2,3,3-Tetrafluoropropoxy)ethoxy]ethan-1-ol
Formula: C₇H₁₂F₄O₃
MW: 220.162 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24842563
CAS Number: 1435806-79-3