1,2-Dibromo-1-(perfluoro-n-hexyl)ethane
Supplier: Apollo Scientific
PC450205-25G PC450205-5G
APOSPC450205-25GEA 503 EUR
APOSPC450205-25G APOSPC450205-5G
1,2-Dibromo-1-(perfluoro-n-hexyl)ethane
1,2-Dibromo-1-(perfluoro-n-hexyl)ethane
Formula: C₈H₃Br₂F₁₃
MW: 505.897 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16621307
CAS Number: 51249-62-8