2-[2-(2,2-Difluoroethoxy)ethoxy]ethan-1-ol
Supplier: Apollo Scientific
Warning
PC450213-250MG PC450213-5G PC450213-1G
APOSPC450213-250MGEA 116 EUR
APOSPC450213-250MG APOSPC450213-5G APOSPC450213-1G
2-[2-(2,2-Difluoroethoxy)ethoxy]ethan-1-ol
2-[2-(2,2-Difluoroethoxy)ethoxy]ethan-1-ol
Formula: C₆H₁₂F₂O₃
MW: 170.154 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24842562
CAS Number: 1435806-96-4