1H,1H,2H,2H,3H,3H,4H,4H-Perfluorodecyl acrylate 95%
Supplier: Apollo Scientific
PC450221-1G
PC450221-5G
APOSPC450221-1GEA
118
EUR
APOSPC450221-1G
APOSPC450221-5G
1H,1H,2H,2H,3H,3H,4H,4H-Perfluorodecyl acrylate 95%
1H,1H,2H,2H,3H,3H,4H,4H-Perfluorodecyl acrylate
Formula:
C₁₃H₁₁F₁₃O₂ MW: 446.204 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18074411 CAS Number: 451456-30-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...