4-Iodo-1,1,1,3-tetrafluoro-2-butene
Supplier: Apollo Scientific
PC450237-1G
PC450237-5G
APOSPC450237-1GEA
190
EUR
APOSPC450237-1G
APOSPC450237-5G
4-Iodo-1,1,1,3-tetrafluoro-2-butene
4-Iodo-1,1,1,3-tetrafluoro-2-butene
Formula:
C₄H₃F₄I MW: 253.965 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08460478 CAS Number: 933668-33-8 |