Tris(2,2,3,3-tetrafluoropropyl)phosphate
Supplier: Apollo Scientific
Danger
PC450238-5G PC450238-1G
APOSPC450238-5GEA 400 EUR
APOSPC450238-5G APOSPC450238-1G
Tris(2,2,3,3-tetrafluoropropyl)phosphate
Tris(2,2,3,3-tetrafluoropropyl)phosphate
Formula: C₉H₉F₁₂O₄P
MW: 440.12 g/mol
Boiling Pt: 140 °C (6 mm Hg)
Melting Pt: –50 °C
Density: 1.6481 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00371442
CAS Number: 563-10-0