1,1,2,2-Tetrafluoroethyl isobutyl ether 99%
Supplier: Apollo Scientific
Danger
PC450241-25G PC450241-5G
APOSPC450241-25GEA 194 EUR
APOSPC450241-25G APOSPC450241-5G
1,1,2,2-Tetrafluoroethyl isobutyl ether 99%
1,1,2,2-Tetrafluoroethyl isobutyl ether
Formula: C₆H₁₀F₄O
MW: 174.137 g/mol
Boiling Pt: 93 °C
Density: 1.1018 g/cm³ (25 °C)
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD16619143
CAS Number: 18180-34-2
UN: 3271
ADR: 3 II