Tris(2,2,3,3,3-pentafluoropropyl)phosphate 97%
Supplier: Apollo Scientific
PC450257-1G PC450257-25G PC450257-5G
APOSPC450257-1GEA 65.5 EUR
APOSPC450257-1G APOSPC450257-25G APOSPC450257-5G
Tris(2,2,3,3,3-pentafluoropropyl)phosphate 97%
Tris(2,2,3,3,3-pentafluoropropyl)phosphate
Formula: C₉H₆F₁₅O₄P
MW: 494.091 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22124059
CAS Number: 25476-41-9