2,2,3,3,5,5,6,6-Octafluoro-1,4-dioxane
Supplier: Apollo Scientific
Warning
PC450276-1G PC450276-5G
APOSPC450276-1GEA 139 EUR
APOSPC450276-1G APOSPC450276-5G
2,2,3,3,5,5,6,6-Octafluoro-1,4-dioxane
2,2,3,3,5,5,6,6-Octafluoro-1,4-dioxane
Formula: C₄F₈O₂
MW: 232.029 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27918494
CAS Number: 32981-22-9