1,3-Bis(1H,1H,3H-tetrafluoropropoxy)-propan-2-ol
Supplier: Apollo Scientific
PC450287-25G PC450287-5G
APOSPC450287-25GEA 919 EUR
APOSPC450287-25G APOSPC450287-5G
1,3-Bis(1H,1H,3H-tetrafluoropropoxy)-propan-2-ol
1,3-Bis(1H,1H,3H-tetrafluoropropoxy)-propan-2-ol
Formula: C₉H₁₂F₈O₃
MW: 320.177 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00427159
CAS Number: 1024024-65-4