3,3,3-Trifluoropropylamine 97%
Supplier: Apollo Scientific
Danger
Synonyms: 3-Amino-1,1,1-trifluoropropane
PC4603-1G PC4603-5G PC4603-500MG
APOSPC4603-1GEA 79.9 EUR
APOSPC4603-1G APOSPC4603-5G APOSPC4603-500MG
3,3,3-Trifluoropropylamine 97%
3,3,3-Trifluoropropylamine
Formula: C₃H₆F₃N
MW: 113.082 g/mol
Boiling Pt: 67…68 °C
Storage Temperature: Ambient
MDL Number: MFCD01694854
CAS Number: 460-39-9
UN: 2924
ADR: 3+8,III