2-(2,2-Difluoroethyl)pyrazol-3-amine
Supplier: Apollo Scientific
PC46351-100MG PC46351-1G PC46351-250MG
APOSPC46351-100MGEA 464 EUR
APOSPC46351-100MG APOSPC46351-1G APOSPC46351-250MG
2-(2,2-Difluoroethyl)pyrazol-3-amine
2-(2,2-Difluoroethyl)pyrazol-3-amine
Formula: C₅H₇F₂N₃
MW: 147.126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09701580
CAS Number: 1171319-69-9