2-Nitro-4-(trifluoromethyl)phenylhydrazine
Supplier: Apollo Scientific
Warning
Synonyms: 4-Hydrazino-3-nitrobenzotrifluoride
PC4673-1G PC4673-250MG
APOSPC4673-1GEA 61.1 EUR
APOSPC4673-1G APOSPC4673-250MG
2-Nitro-4-(trifluoromethyl)phenylhydrazine
2-Nitro-4-(trifluoromethyl)phenylhydrazine
Formula: C₇H₆F₃N₃O₂
MW: 221.137 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00622442
CAS Number: 1513-50-4