2,3-Dihydro-8-fluoroquinolin-4(1H)-one 97%
Supplier: Apollo Scientific
Danger
Synonyms: 8-Fluoro-4-oxo-1,2,3,4-tetrahydroquinoline
PC4676-1G PC4676-250MG
APOSPC4676-1GEA 345 EUR
APOSPC4676-1G APOSPC4676-250MG
2,3-Dihydro-8-fluoroquinolin-4(1H)-one 97%
2,3-Dihydro-8-fluoroquinolin-4(1H)-one
Formula: C₉H₈FNO
MW: 165.164 g/mol
Melting Pt: 125…127 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD07784308
CAS Number: 38470-28-9
UN: 2811
ADR: 6.1 III