1,3-Bis(tridecafluorohexyl)benzene 97%
Supplier: Apollo Scientific
Synonyms: 1,3-Bis(1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorohexyl)benzene
PC47465-1G PC47465-5G
APOSPC47465-1GEA 115 EUR
APOSPC47465-1G APOSPC47465-5G
1,3-Bis(tridecafluorohexyl)benzene 97%
1,3-Bis(tridecafluorohexyl)benzene
Formula: C₁₈H₄F₂₆
MW: 714.183 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27918468
CAS Number: 124389-29-3