3-(Difluoromethoxy)-4-fluorobromobenzene
Supplier: Apollo Scientific
Synonyms: 5-Bromo-α,α,2-trifluoroanisole, 4-Bromo-2-(1,1-difluoromethoxy)-1-fluorobenzene, 4-Bromo-2-(difluoromethoxy)-1-fluorobenzene
PC48299-25G PC48299-5G
APOSPC48299-25GEA 823 EUR
APOSPC48299-25G APOSPC48299-5G
3-(Difluoromethoxy)-4-fluorobromobenzene
3-(Difluoromethoxy)-4-fluorobromobenzene
Formula: C₇H₄BrF₃O
MW: 241.005 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11036068
CAS Number: 954235-78-0