4-(Trifluoromethyl)pyridazine 97%
Supplier: Apollo Scientific
Danger
Synonyms: 4-(Trifluoromethyl)-1,2-diazine
PC48363-1G PC48363-500MG PC48363-5G
APOSPC48363-1GEA 263 EUR
APOSPC48363-1G APOSPC48363-500MG APOSPC48363-5G
4-(Trifluoromethyl)pyridazine 97%
4-(Trifluoromethyl)pyridazine
Formula: C₅H₃F₃N₂
MW: 148.086 g/mol
Density: 1.4 g/cm³ (25 °C)
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD11226564
CAS Number: 132537-44-1
UN: 2810
ADR: 6.1,III