Methyl 4-methoxy-2-(trifluoromethyl)benzoate
Supplier: Apollo Scientific
Warning
Synonyms: 5-Methoxy-2-(methoxycarbonyl)benzotrifluoride, 4-(Methoxycarbonyl)-3-(trifluoromethyl)anisole
PC48410-1G PC48410-5G PC48410-25G
APOSPC48410-1GEA 29 EUR
APOSPC48410-1G APOSPC48410-5G APOSPC48410-25G
Methyl 4-methoxy-2-(trifluoromethyl)benzoate
Methyl 4-methoxy-2-(trifluoromethyl)benzoate
Formula: C₁₀H₉F₃O₃
MW: 234.172 g/mol
Melting Pt: 47…48 °C
Storage Temperature: Ambient
MDL Number: MFCD06204092
CAS Number: 773875-67-5