4-Bromo-3-chloro-2-fluorobenzaldehyde 97%
Supplier: Apollo Scientific
Warning
PC48449-1G PC48449-250MG PC48449-5G
APOSPC48449-1GEA 313 EUR
APOSPC48449-1G APOSPC48449-250MG APOSPC48449-5G
4-Bromo-3-chloro-2-fluorobenzaldehyde 97%
4-Bromo-3-chloro-2-fluorobenzaldehyde
Formula: C₇H₃BrClFO
MW: 237.454 g/mol
Melting Pt: 77…79 °C
Storage Temperature: Ambient
MDL Number: MFCD26958726
CAS Number: 1696224-75-5