3-Fluoro-4-iodo-2-methylaniline
Supplier: Apollo Scientific
Warning
PC48454-1G PC48454-250MG PC48454-5G
APOSPC48454-1GEA 163 EUR
APOSPC48454-1G APOSPC48454-250MG APOSPC48454-5G
3-Fluoro-4-iodo-2-methylaniline
3-Fluoro-4-iodo-2-methylaniline
Formula: C₇H₇FIN
MW: 251.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21364950
CAS Number: 329927-16-4