3-Difluoromethyl-5-(4-methoxyphenyl)pyrazole 97%
Supplier: Apollo Scientific
Warning
PC4849-1G PC4849-5G
APOSPC4849-1GEA 87 EUR
APOSPC4849-1G APOSPC4849-5G
3-Difluoromethyl-5-(4-methoxyphenyl)pyrazole 97%
3-Difluoromethyl-5-(4-methoxyphenyl)pyrazole
Formula: C₁₁H₁₀F₂N₂O
MW: 224.207 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04039282
CAS Number: 1029636-42-7