(2S)-3,3,3-Trifluoropropane-1,2-diol
Supplier: Apollo Scientific
Warning
PC48579-1G PC48579-250MG
APOSPC48579-1GEA 1680 EUR
APOSPC48579-1G APOSPC48579-250MG
(2S)-3,3,3-Trifluoropropane-1,2-diol
(2S)-3,3,3-Trifluoropropane-1,2-diol
Formula: C₃H₅F₃O₂
MW: 130.066 g/mol
Boiling Pt: 87…88 °C (25 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD22199791
CAS Number: 148683-14-1
UN: 1993
ADR: 3,III