2,2',2''-(2,4,6-Trifluorobenzene-1,3,5-triyl)tris(4,4,5,5-tetramethyl-1,3,2-dioxaborolane)
Supplier: Apollo Scientific
Warning
PC48621-1G PC48621-5G
APOSPC48621-1GEA 430 EUR
APOSPC48621-1G APOSPC48621-5G
2,2',2''-(2,4,6-Trifluorobenzene-1,3,5-triyl)tris(4,4,5,5-tetramethyl-1,3,2-dioxaborolane)
2,2',2''-(2,4,6-Trifluorobenzene-1,3,5-triyl)tris(4,4,5,5-tetramethyl-1,3,2-dioxaborolane)
Formula: C₂₄H₃₆B₃F₃O₆
MW: 509.967 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12407207
CAS Number: 408492-24-0