4-[(Difluoromethyl)sulphonyl]bromobenzene
Supplier: Apollo Scientific
Warning
PC48638-1G PC48638-250MG
APOSPC48638-1GEA 382 EUR
APOSPC48638-1G APOSPC48638-250MG
4-[(Difluoromethyl)sulphonyl]bromobenzene
4-[(Difluoromethyl)sulphonyl]bromobenzene
Formula: C₇H₅BrF₂O₂S
MW: 271.079 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13188442
CAS Number: 51679-57-3