Methyl 3-amino-7-(trifluoromethyl)-1-benzothiophene-2-carboxylate
Supplier: Apollo Scientific
Warning
PC48670-1G PC48670-250MG
APOSPC48670-1GEA 208 EUR
APOSPC48670-1G APOSPC48670-250MG
Methyl 3-amino-7-(trifluoromethyl)-1-benzothiophene-2-carboxylate
Methyl 3-amino-7-(trifluoromethyl)-1-benzothiophene-2-carboxylate
Formula: C₁₁H₈F₃NO₂S
MW: 275.247 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29169513
CAS Number: 2379918-54-2