8-Fluoroquinoline-3-carboxylic acid
Supplier: Apollo Scientific
Warning
PC49033-1G PC49033-250MG PC49033-5G
APOSPC49033-1GEA 62.5 EUR
APOSPC49033-1G APOSPC49033-250MG APOSPC49033-5G
8-Fluoroquinoline-3-carboxylic acid
8-Fluoroquinoline-3-carboxylic acid
Formula: C₁₀H₆FNO₂
Melting Pt: 240…250 °C
Storage Temperature: Ambient
MDL Number: MFCD11108738
CAS Number: 71082-53-6