2-Chloroethyl trifluoromethanesulphonate 95%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Chloroethyl triflate
PC49047-1G PC49047-5G
APOSPC49047-1GEA 284 EUR
APOSPC49047-1G APOSPC49047-5G
2-Chloroethyl trifluoromethanesulphonate 95%
2-Chloroethyl trifluoromethanesulphonate
Formula: C₃H₄ClF₃O₃S
MW: 212.575 g/mol
Melting Pt: –50.5…–50 °C
Density: 1.6 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD18074401
CAS Number: 73323-80-5
UN: 2810
ADR: 6.1 III