2,3-Difluoro-4-iodoaniline
Supplier: Apollo Scientific
Warning
PC49086-1G PC49086-5G
APOSPC49086-1GEA 87 EUR
APOSPC49086-1G APOSPC49086-5G
2,3-Difluoro-4-iodoaniline
2,3-Difluoro-4-iodoaniline
Formula: C₆H₄F₂IN
MW: 255.004 g/mol
Melting Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD21602052
CAS Number: 1026989-94-5