1-[3-Chloro-5-(trifluoromethyl)pyridin-2-ylamino]-2-[2-amino-4-(trifluoromethyl)anilino]ethane 97%
Supplier: Apollo Scientific
PC4918-1G PC4918-500MG
APOSPC4918-1GEA 799 EUR
APOSPC4918-1G APOSPC4918-500MG
1-[3-Chloro-5-(trifluoromethyl)pyridin-2-ylamino]-2-[2-amino-4-(trifluoromethyl)anilino]ethane 97%
1-[3-Chloro-5-(trifluoromethyl)pyridin-2-ylamino]-2-[2-amino-4-(trifluoromethyl)anilino]ethane
Formula: C₁₅H₁₃ClF₆N₄
MW: 398.734 g/mol
Melting Pt: 143…145 °C
Storage Temperature: Ambient
MDL Number: MFCD00793113
CAS Number: 478262-14-5