4-(cis-3,5-Dimethylpiperazin-1-yl)-3-fluoroaniline
Supplier: Apollo Scientific
Warning
Synonyms: 1-(4-Amino-2-fluorophenyl)-cis-3,5-dimethylpiperazine
PC49206-1G PC49206-250MG
APOSPC49206-1GEA 325 EUR
APOSPC49206-1G APOSPC49206-250MG
4-(cis-3,5-Dimethylpiperazin-1-yl)-3-fluoroaniline
4-(cis-3,5-Dimethylpiperazin-1-yl)-3-fluoroaniline
Formula: C₁₂H₁₈FN₃
MW: 223.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13619678
CAS Number: 943752-32-7