5'-Bromo-2'-fluoro-3'-nitroacetanilide
Supplier: Apollo Scientific
Warning
Synonyms: N-(5-Bromo-2-fluoro-3-nitrophenyl)acetamide, 3-Acetamido-5-bromo-2-fluoronitrobenzene
PC49219-1G PC49219-250MG
APOSPC49219-1GEA 1320 EUR
APOSPC49219-1G APOSPC49219-250MG
5'-Bromo-2'-fluoro-3'-nitroacetanilide
5'-Bromo-2'-fluoro-3'-nitroacetanilide
Formula: C₈H₆BrFN₂O₃
MW: 277.047 g/mol
Melting Pt: 175 °C
Storage Temperature: Ambient
MDL Number: MFCD19441790
CAS Number: 1313588-91-8