4-(Trifluoromethoxy)phenoxyacetyl chloride
Supplier: Apollo Scientific
Danger
PC4923-1G PC4923-5G
APOSPC4923-1GEA 194 EUR
APOSPC4923-1G APOSPC4923-5G
4-(Trifluoromethoxy)phenoxyacetyl chloride
4-(Trifluoromethoxy)phenoxyacetyl chloride
Formula: C₉H₆ClF₃O₃
MW: 254.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02091505
CAS Number: 72220-51-0
UN: 1760
ADR: 8 III