5-[1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-yl]thiophene-2-carboxylic acid
Supplier: Apollo Scientific
Warning
Synonyms: 3-(5-Carboxythien-2-yl)-1-methyl-5-(trifluoromethyl)-1H-pyrazole
PC4924-100MG PC4924-1G
APOSPC4924-100MGEA 72.5 EUR
APOSPC4924-100MG APOSPC4924-1G
5-[1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-yl]thiophene-2-carboxylic acid
5-[1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-yl]thiophene-2-carboxylic acid
Formula: C₁₀H₇F₃N₂O₂S
MW: 276.235 g/mol
Melting Pt: 212…215 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00206674
CAS Number: 223499-20-5