3-Hydroxy-5-(trifluoromethyl)benzaldehyde
Supplier: Apollo Scientific
Warning
Synonyms: 3-Formyl-5-hydroxybenzotrifluoride, 3-Formyl-5-(trifluoromethyl)phenol
PC49281-1G PC49281-250MG PC49281-5G
APOSPC49281-1GEA 145 EUR
APOSPC49281-1G APOSPC49281-250MG APOSPC49281-5G
3-Hydroxy-5-(trifluoromethyl)benzaldehyde
3-Hydroxy-5-(trifluoromethyl)benzaldehyde
Formula: C₈H₅F₃O₂
MW: 190.119 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16999720
CAS Number: 1243312-51-7