2-(Trifluoromethyl)isonicotinonitrile ≥95%
Supplier: Apollo Scientific
Warning
Synonyms: 2-(Trifluoromethyl)pyridine-4-carbonitrile
PC49309-250MG PC49309-1G
APOSPC49309-250MGEA 174 EUR
APOSPC49309-250MG APOSPC49309-1G
2-(Trifluoromethyl)isonicotinonitrile ≥95%
2-(Trifluoromethyl)isonicotinonitrile
Formula: C₇H₃F₃N₂
MW: 172.107 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11848100
CAS Number: 916210-02-1